If you want to save a custom filter to a configuration (.cfg) file for future use, follow the procedure below.
Procedure
  1. In the Results table on the Process and Review Page, right-click anywhere in the table and select Filters > Save As from the shortcut menu.
  2. The Save As dialog box opens.
  3. Browse to the folder location on your computer where you want to save the custom filter file.
  4. Save the file with the ".cfg" extension.
  5. The saved configuration file retains all of the current filters at the top level (component level) in the table.