The following table lists the various filter operators for the different types of data in the table columns.
Type of data | Filter operator | Condition description |
|---|---|---|
Textual and Numerical | Equals | Data is equal to the selected filter operand or a value that you enter in the operand box. |
Not equals | Data is not equal to the selected filter operand or a value that you enter in the operand box. | |
Less than | Data is less than the selected filter operand or a value that you enter in the operand box. | |
Less than or equal to | Data is less than or equal to the selected filter operand or a value that you enter in the operand box. | |
Greater than | Data is greater than the selected filter operand or a value that you enter in the operand box. | |
Greater than or equal to | Data is greater than the selected filter operand or a value that you enter in the operand box. | |
In | Data contains the selected filter operand or a substring that you enter in the operand box. | |
Not in | Data does not contain the selected filter operand or a substring that you enter in the operand box. | |
Textual | Contains | Data contains the selected filter operand or text that you enter in the operand box. |
Does not contain | Data does not contain the selected filter operand or text that you enter in the operand box. | |
Like (wildcards) | Data is like the selected filter operand or text and a wildcard character that you enter in the operand box. Use these wildcard characters:
| |
Not like (wildcards) | Data are not like the selected filter operand or text and a wildcard character that you enter in the operand box. Use these wildcard characters:
| |
Match (regular expression) | Data matches a regular expression that you enter in the operand box. | |
Does not match (regular expression) | Data does not match a regular expression that you enter in the operand box. | |
Starts with | Data starts with the selected filter operand or text that you enter in the operand box. | |
Does not start with | Data does not start with the selected filter operand or text that you enter in the operand box. | |
Ends with | Data ends with the selected filter operand or text that you enter in the operand box. | |
Does not end with | Data does not end with the selected filter operand or text that you enter in the operand box. | |
Numerical | Top | Enter a number, n, in the operand box. The filter column displays only the highest n numbers out of all the numbers in that column. |
Bottom | Enter a number, m, in the operand box. The filter column displays only the lowest m numbers out of all the numbers in that column. | |
Top Percentile | Enter a number, x, in the operand box. The filter column displays only the highest x% out of all the numbers in that column. | |
Bottom Percentile | Enter a number, y, in the operand box. The filter column displays only the lowest y% out of all the numbers in that column. |