When you launch the BioPharma Finder 5.2 application for the first time, it opens by default in the local mode, disconnected from the Ardia platform. The home page displays the Experiment Types navigation pane and workflow links.
BioPharma Finder home page (local mode)
BioPharma Finder home page (local mode)
The Experiment Types navigation pane contains five tabs that open the Sequence Manager or any of the following analyses included in the BioPharma Finder application:
  • Peptide Mapping Analysis
  • Oligonucleotide Analysis
  • Intact Mass Analysis
  • Top Down Analysis
The type of license you purchased and set during the installation controls which analysis workflows are active. You can start the Sequence Manager and any activated analysis workflow by selecting the corresponding tab.