Import a workbook - Import a workbook - BioPharma Finder 5.2 Oligonucleotide Analysis User Guide - BioPharma Finder Software - Import a workbook - BioPharma Finder Software - BioPharma Finder Software - topic
BioPharma Finder 5.2 Oligonucleotide Analysis User Guide
- Product Tree
- Ardia Platform > Software > BioPharma Finder Software
- Desktop Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- On the BioPharma Finder home page, select the Oligonucleotide Analysis workflow link in the Experiment Types pane or below the splash graphic.
- The Oligonucleotide Analysis page opens.
- Select the Calculator and Workbook tab on the navigation bar.
- If necessary, select the Workbook Manager subtab.
- The Workbook Manager page opens, showing the saved workbooks in a table.
- To import one or more workbooks, select Import on the command bar.
- A dialog box opens showing the default file location:
C:\Xcalibur\data. - You can browse to another folder as necessary.
- Select the workbooks you want to import and then select OK.
- The application adds the workbooks to the Workbook Manager table.
- If the imported workbook has the same name as an existing workbook, the applications appends _imported to the file name of the new workbook.