The following table describes the commands and parameters in the Oligonucleotide Calculator and Workbook Editor.
Parameter | Description |
|---|---|
Command bar | |
Import Sequence | Imports multiple oligonucleotide sequences at once from a specially-formatted .csv file. |
Export | Exports the currently open oligo workbook as a Microsoft™ Excel™ file (.xslx) or oligo workbook (.wbpf). |
Save | Saves the oligo workbook to the Workbook Manager. |
Close | Closes the Oligonucleotide Calculator and Workbook Editor. |
Manual Sequence subtab | |
Chain Name | Specifies the name of an added chain in the oligonucleotide sequence. |
(Editor box) | Provides an area for you to manually type or paste new chain information. |
Apply (button) (left side of page) | Adds the new chain to the Results table. |
Apply to All Sequences (checkbox) | Specifies that changes to the parameters will be applied to all sequences in the Results table. |
Apply to Selected Sequences (checkbox) | Specifies that changes to the parameters will be applied only to the sequences currently selected in the Results table. |
Charge State Range | Specifies the range of charge states of the sequences in the Results table. Charge state refers to the imbalance between the number of protons and the number of electrons in a molecular species (or an ion adduct). Default values: −20 to −2 Range of values: −100 to 100 Note: The minimum and maximum charge state values must be of the same polarity (both values negative or positive). |
m/z Range | Specifies the range of the mass-to-charge ratios of the sequences in the Results table. Default values: 300 to 2500 Range of values: 10 to 100,000 |
Mass Range (M) | Specifies the range of masses of the sequences in the Results table. Default values: 200 to 65,000 Range of values: 10 to 100,000 |
Number of Isotopes | Specifies the number of isotopes to include for each sequence in the Results table, based on the most abundant isotope per oligonucleotide. Range of values: 0 to 15 |
Optional Columns (checkboxes) | (Selected by default) Select the checkbox to include columns listing the monoisotopic masses of several possible adducts and base losses in the Results table. Adduct columns include:
Base loss (M−B) columns include:
|
Apply (button) (right side of page) | Updates the Results table with the parameters specified for all or selected sequences. |
Modification subtab | |
Apply to All Sequences (checkbox) | Specifies that the modifications will be applied to all sequences in the Results table. |
Apply to Selected Sequences (checkbox) | Specifies that the modifications will be applied only to the sequences currently selected in the Results table. |
Update (button) (left side of page) | Updates the sequences in the Results table with the specified global subunit modifications. |
Subunit/From/To (drop-down lists) | Specifies global changes to a subunit from one group to another. |
5' Terminal (drop-down list) | Specifies the 5'-terminal group for a selected triplet. |
(Subunit table) | Specifies subunit changes for a selected triplet. |
3' Terminal (drop down list) | Specifies the 3'-terminal group for a selected triplet. |
Update (button) (right side of page) | Updates the sequences in the Results table with the specified triplet-specific subunit modifications. |
Add/Edit Building Blocks (subtab) | |
(Table of building blocks) | Lists all of the individual subunits and their properties (name, symbol/site, formula, monoisotopic mass, and average mass). |
Edit (button) | Select to modify the properties of the subunit selected in the table of building blocks. |
Delete (button) | Select to delete the subunit selected in the table of building blocks. |
Oligo building block properties (section) | Lists the editable properties of the subunit selected in the table of building blocks. You can edit the name, symbol, and formula for each subunit. The monoisotopic mass and average mass fields are read only. Note: You cannot edit any variable modifications or delete default subunits. |
Apply (button) | Adds/updates the edited oligo building block in the table of building blocks. |