The following table describes the format of the oligonucleotide modification information listed in the "Mod" column in the Results tables on the Process and Review page.
Modification notation information
Modification type
Description
Variable modification
For example, 3:A9–T15 = 2104.4033M(A9+Deamination) shows:
  • A9 = the modification occurs at the adenine at position 9 in oligonucleotide 3
  • Deamination = the modification type
Multiple modifications
For example, 7G1–C15 = 4671.7648m[3’p](C6+C Depyrimidination) shows two modifications:
  • A phosphate group (p) at the 3’ terminal (C15 end) of the sequence 7G1–C15 in oligonucleotide 7
  • A depyrimidination at base C6 in the sequence 7G1–C15 in oligonucleotide 7
Unspecified modification
An unspecified modification is the addition or subtraction of a given mass of unspecified origin.
For example, 6:A1–G17 = 5159.9221m(~C10+14.0137) shows:
  • ~C10 = the modification is on the C base at position 10 (approximately) in oligonucleotide 6
  • +14.0137= the addition of a mass of 14.0137 amu
The Enable Mass Search for Unspecified Modifications option must be selected in the processing method (Parameters > Identification > Advanced Search) for the application to perform these identifications.
Nonspecific
For example, 1:A3–T17 = 4595.034m[nonspecific] shows that the identified oligonucleotide was produced from an unexpected cleavage for the specified ribonuclease.
3’-terminal modifications
The 3’-terminal of an oligonucleotide can be modified with the following groups:
  • Phosphate (p): H3PO4
  • Phosphorothioate (s): H3PO3S
  • 3’-Triantennery GalNAc (g): C78H140N11O34P
For example, 7:G10–C21 = 3726.378m[3’p] shows that the 3’-terminal (C21 end) of the sequence G10–C21 of oligonucleotide 7 has been modified with a phosphate (p) group.
5’-terminal modifications
The 5’-terminal of an oligonucleotide can be modified with the following groups:
  • Phosphate (p): H3PO4
  • Phosphorothioate (s): H3PO3S
  • Biotin (b): C10H16N2O3S
  • cAG (a): C32H44O27N15P5
  • cAU (u): C31H43O28N12P5
  • cARCA (r): C22H32O21N10P4
  • mCAP (c): C21H30O21N10P4
For example, 1:A26-G28 = 997.15180m[5'p] shows that the 5’-terminal (A26 end) of the sequence A26–G28 of oligonucleotide 1 has been modified with a phosphate (p) group.
Adducts
For example, 8:T1–G24 = 7342.2761m(Na+) shows that the oligonucleotide is a Na+ adduct.
The application detects adducts when an identified mass is higher than the expected precursor mass by a specific amount, for example, ~22 amu higher for a Na+ adduct, based on the given charge state.
The application search for many possible adducts, including Na+, 2Na+, Ca+2, and Fe+3.
Deletion
A deletion (residual loss) is the loss of a nucleotide base within a given sequence identification.
The Enable Residue Deletion option must be selected in the processing method (Parameters > Identification > Advanced Search) for the application to perform these identifications.
The “Site” column in the Results table displays the deleted nucleotide base.
For example, 1:U1–G6 = 1897.3107m(G4+Deletion) shows that the application detected that nucleotide base G4 was deleted from the sequence U1–G6 in oligonucleotide 1. The Site column displays “G4”.
NOTE
A tilde (~) displayed before an amino acid site (~G11) indicates that the position of the identified modification is approximate (less definite).