You can specify which components to include in the Oligonucleotide Sequence Coverage Map on the Coverage page by selecting specific components in the Results table on the Process and Review page.
Prerequisites
- You have loaded the experiment results are viewing the Coverage page under the Mapping tab.
Procedure
- From the Coverage page, select the Process and Review tab in the navigation bar.
- In the Results table, select or clear the checkboxes for the components that you want to include in or exclude from the Oligonucleotide Sequence Coverage Map on the Coverage page.
- Navigate back to the Oligonucleotide Sequence Coverage Map on the Coverage page by selecting the Mapping > Coverage tab in the navigation bar.
- The Oligonucleotide Sequence Coverage Map now shows only the components selected on the Process and Review page.
- The map title now displays "Oligonucleotide Sequence Coverage MapāUser Defined".
- To return to the default Oligonucleotide Sequence Coverage Map, repeat the process above, but in the Results table on the Process and Review page, select the checkbox in the table header row to clear all of the checkboxes and deselect all components.