Right-clicking the MS2 Spectra pane on the Process and Review page opens a shortcut menu with the commands listed in the following table.
Command | Description |
|---|---|
Reset Scale | Restores the original scale to the MS2 Spectra pane. |
Copy | Copies the image viewable in the MS2 Spectra pane, including labeling and shading. |
Predict Oligo MS/MS (Kinetic Model) | Uses the Kinetic model to generate the predicted spectrum in the MS2 Spectra pane. |
Delete Predicted MS/MS Spectrum | Deletes the predicted spectrum in the MS2 Spectra pane. To redisplay the predicted spectrum, you can either reselect the component row in the Results table or right-click in the MS2 Spectra pane and then select the Predict Oligo MS/MS (Kinetic Model) command from the shortcut menu. |