In the MS2 Spectra pane, the predicted MS/MS spectrum is displayed on the top of the experimental spectrum by default. You can change this view to compare the experimental spectra of two different components.
Prerequisites
- You have loaded the experiment results and are viewing the MS2 Spectra pane on the Process and Review page.
Procedure
- In the Results table, select the row of a component that has an MS/MS spectrum.
TIP
In the Results table, filter the ID Type column to "MS2" to display the components with an associated MS/MS spectrum.
- The MS2 Spectra pane view displays the predicted spectrum on the top and the experimental spectrum on the bottom for the selected component.
- In the MS2 Spectra pane, right-click anywhere on the spectra.
- In the shortcut menu, the "Predicted view" checkbox is selected by default.
- Select the "Set bottom spectrum as a reference (move to top)" option in the shortcut menu.
- The "Predicted view" checkbox is cleared.
- The experimental MS/MS spectrum on the bottom moves to the top view and replaces the predicted spectrum. This experimental spectrum is now set as the reference spectrum.
- In the Results table, select the row of a different component that has an MS/MS spectrum you want to compare.
- The MS2 Spectra pane displays the stacked spectra for comparison with the reference spectrum displayed on the top.
Comparing the MS/MS spectra of two components, shown for a peptide mapping analysis experiment NOTE
The experimental reference spectrum that you set in the MS2 Spectra pane is not linked between pages in the application.
That is, when you set a component spectrum as the reference on the Process and Review page, this information is not passed to the Mapping page.
- When comparing two experimental spectra, the Predict Peptide MS/MS (Kinetic Model) functionality is deactivated on the shortcut menu.
- The experimental reference spectrum remains in its location on top until you right-click and select the "Predicted view" checkbox to restore the default view.
- When you select "Predicted view", the experimental reference spectrum is replaced with the predicted spectrum of the component whose experimental spectrum is on the bottom.