You can export the information at all levels in the Coverage Results table as a Microsoft™ Excel™ worksheet (.xlsx).
Prerequisites
- You have loaded the experiment results are viewing the Coverage page under the Mapping tab.
Procedure
- On the Coverage page, right-click anywhere in the Coverage Results table and select Export in the shortcut menu.
- The Save As dialog box opens.
- Browse to the file location where you want to save your results and type a file name.
- By default, the application navigates to the folder containing the raw data files for the experiment.
- By default, the file name is the same name as the experiment.
- Select Save.
- The application saves the results as a Microsoft™ Excel™ worksheet (.xlsx).
- The file contains the results at both the oligonucleotide and raw data file levels (if applicable) for all oligonucleotides listed in the Results table.