The Identification page includes three sections: Oligonucleotide Identification, Select Ribonuclease, and Advanced Search.
Parameter | Description |
|---|---|
Oligonucleotide Identification | |
Search by Full MS Only | Indicates the type of data that application will use to search.
|
Use MS/MS | Specifies the type of data to process.
|
Maximum Oligonucleotide Mass | Specifies the maximum oligonucleotide mass to be identified.
|
Mass Accuracy (ppm) | Specifies the maximum deviation (ppm) between the mass of a theoretical oligonucleotide and the calculated mass of a particular ion used to identify an oligonucleotide
|
Minimum Confidence | Specifies the minimum confidence level for a reported oligonucleotide assignment on a 0-to-1 scale, with 1 being the highest confidence.
If “Yes” is selected in the Search by Full MS Only field, the minimum confidence will be set to 0.00. |
Maximum Number of Modifications for an Oligonucleotide | Read-only. Specifies the maximum number of modifications allowed for each oligonucleotide; previously specified in the Assign Variable Modifications area of the Sequence Editor page. See Manage variable modifications. For oligonucleotide analysis, only one modification for each oligonucleotide identified is supported at this time.
|
Advanced Search | |
Enable Mass Search for Unspecified Modifications (checkbox) | Select the Enable Mass Search for Unspecified Modifications checkbox to allow the application to perform a mass search for unspecified modifications to the oligonucleotide. |
Mass Changes for Unspecified Modifications | Defines the range of changes to the mass of a nucleotide resulting from an unspecified modification. Only integer values are possible.
Read-only if the Enable Mass Search for Unspecified Modifications checkbox is not selected. |
Enable Residue Deletion (checkbox) | Select the Enable Residue Deletion checkbox to allow the application to search for possible oligonucleotide sequences that may be missing a single nucleotide at any position. |
Select Ribonuclease | |
RNase (drop-down menu) | Specifies the ribonuclease assigned to the currently open processing method.
Ribonuclease notation information: A dash (-) indicates the cutting position of the indicated ribonuclease. For example, “-ACA” indicates a cut before the ACA sequence, “G-” indicates a cut after G bases, and “G-U” indicates a cut between G and U bases. |
Custom Specificity (checkbox) | Specifies any custom specificity applied to the ribonuclease assigned to the currently open processing method. By default, the Custom Specificity field is inactive (gray) but you can enable it for the selected RNase by selecting the Custom Specificity checkbox.
|
Phosphate Location | Specifies the phosphate location defined for the currently open processing method.
|
Specificity Level | Defines the level of ribonuclease specificity for the ribonuclease assigned to the currently open processing method.
Specificity levels High, Medium, and Low correspond to confidence factors that the application uses to determine the final confidence score of component identification. The difference between these levels can impact your results when two or more components match the spectrum, but only one of the components adheres to the specificity requirements of the selected ribonuclease. |