The following table describes the parameters in the Assign Variable Modifications pane in the Oligonucleotide Sequence Editor.
Parameter | Description |
|---|---|
Max # Modifications | Specifies the maximum number of variable modifications that you want to assign to the target oligonucleotide sequence. Range: 0 to 8; default: 1 |
Modifications | Lists the modifications available to apply to the 5'-terminal, 3'-terminal, and oligonucleotide building blocks. |
5'-Terminal | Displays information about the 5'-terminal modification that you selected from the list. |
Read-only fields | |
Mono. Mass | Displays the monoisotopic mass of the selected 5'-terminal modification to three decimal places. |
Avg. Mass | Displays the average mass of the selected 5'-terminal modification to two decimal places. |
Buttons | |
Add | Adds the selected modification to the Modifications Selected for Search list. |
Remove | Removes the selected modification from the Modifications Selected for Search list. |
Load Default Mods | Quickly loads the sublist of default modifications into the Modifications Selected for Search list. |
3'-Terminal | Displays information about the 3'-terminal modification that you selected from the list. |
Read-only fields | |
Mono. Mass | Displays the monoisotopic mass of the selected 3'-terminal modification to three decimal places. |
Avg. Mass | Displays the average mass of the selected 3'-terminal modification to two decimal places. |
Buttons | |
Add | Adds the selected modification to the Modifications Selected for Search list. |
Remove | Removes the selected modification from the Modifications Selected for Search list. |
Load Default Mods | Quickly loads the sublist of default modifications into the Modifications Selected for Search list. |
Oligo Variable Modifications | Displays information about the oligo variable modification that you selected from the list. |
Read-only fields | |
Mono. Mass | Displays the monoisotopic mass of the selected oligo variable modification to three decimal places. |
Avg. Mass | Displays the average mass of the selected oligo variable modification to two decimal places. |
Residues | Displays residue for the selected oligo variable modification. |
Buttons | |
Add | Adds the selected modification to the Modifications Selected for Search list. |
Remove | Removes the selected modification from the Modifications Selected for Search list. |
Load Default Mods | Quickly loads the sublist of default modifications into the Modifications Selected for Search list. |