Procedure
- In the Oligonucleotide Sequence Editor, select the title bar for the Building Block and Variable Modification Editor.
- The Building Block and Modification Editor pane opens, displaying the Oligo Building Block and Oligo Variable Modification areas.
- For more information on creating custom variable modifications, see Add custom modifications.
- In the Oligo Building Block area, select the subunit for the type of custom building block you want to create from the Subunit list.
- The subunit types you can customize include base, backbone linker, 2'-ribose, and 3'- and 5'-terminal.
- In the Name box, enter a unique name for the custom building block.
- In the Symbol box, select a symbol to denote the custom building block.
NOTE
For a custom base, the application does not accept the following symbols: A, G, U, T, C, I, or H. These symbols appear in the list, but are reserved for the default bases.
- In the formula box, enter the chemical formula of the custom building block.
- The application only recognizes the symbols for the elements in the periodic table and integers for the number of atoms.
- For the base, backbone linker, and 2' ribose subunits, the application supports only these elements: C, H, O, N, S, P, and F.
- To define the loss of a 2'-ribose group as custom building block, type a hyphen (-) at the beginning of the formula.
- The application does not accept the loss of bases, backbone linkers, the 5'-terminal, or the 3'-terminal as a custom building block.
- Enter the monoisotopic mass and the average mass for the modification in the respective fields.
- If your modification involves a loss of mass, enter negative monoisotopic and average mass values.
- Select Apply.
- The application saves the custom building block and the modification appears in purple in the modification list at the bottom of the pane.
- Repeat steps 2 through 7 to create additional custom building blocks as necessary.
- Examples of custom oligo building blocks are shown in the figures below.
Custom 3'-terminal (N-acetylgalactosamine) created in the Oligonucleotide Sequence Editor Custom 5'-terminal (hydroxide) created in the Oligonucleotide Sequence Editor Custom backbone linker (thiol) created in the Oligonucleotide Sequence Editor