The following table lists the various filter operands for the different types of data in the table columns.
Type of data | Filter operand | Operand description |
---|---|---|
Textual | Custom | Set up groups of multiple filter conditions. |
Blanks | Blank value | |
NonBlanks | Not a blank value | |
Text | A specific text string | |
Numerical | Custom | Set up groups of multiple filter conditions. |
Blanks | Blank value | |
NonBlanks | Not a blank value | |
Number | A specific number |