You can zoom in/out on, reset, and copy the spectra in the application.
Zoom in or zoom out
Procedure
- To zoom in on a spectrum, do either of the following:
- In the spectrum pane, drag the red cross-shaped cursor over the peak or peaks of interest to form a box.
Enlarging a peak by drawing a box around it - In the spectrum pane, hold down the left mouse button and draw a line beneath the baseline of the peaks of interest.
Enlarging a peak by drawing a line beneath its baseline - When you zoom in on a region of the spectrum, the application recalculates the values on the y axis so that 100 percent represents the highest intensity in the displayed region. The intensities shown on the y axis remain at values relative to the most intense component in the plot.
- To zoom out on a spectrum, right-click the spectrum pane and select Zoom Out (if available). Otherwise, reset the spectrum view to the original scale zoom out (see below).
Reset spectrum view to original scale
Procedure
- To reset the spectrum view to the original scale, do either of the following:
- Right-click the spectrum pane and select Reset Scale.
- Double-click anywhere in the spectrum pane.
Copy spectrum to the clipboard
Procedure
- To copy a spectrum to the clipboard, right-click the spectrum pane and select Copy, Copy as Displayed, or Copy per Global Settings.
- The application copies the entire view of the spectrum pane to the clipboard, including all visible labeling and shading. You can then paste the copied picture into a third-party application file.
- See Copy and paste functions.