You can zoom in/out on, reset, copy, and label the chromatograms in the application.
Zoom in or out
Procedure
- To zoom in on a chromatogram, do either of the following:
- In the Chromatogram pane, drag the red cross-shaped cursor over the peak or peaks of interest to form a box.
- Note: For intact mass analysis or top down analysis, first select the Auto Zooming option on the pane title bar to activate the zooming command.
Enlarging a peak by drawing a box around it - In the Chromatogram pane, hold down the left mouse button and draw a line beneath the baseline of the peaks of interest.
- Note: For intact mass analysis or top down analysis, first select the Auto Zooming option on the pane title bar to activate the zooming command.
Enlarging a peak by drawing a line beneath its baseline - When you zoom in on a region of the chromatogram, the application recalculates the values on the y axis so that 100 percent represents the highest intensity in the displayed region. The intensities shown on the y axis remain at values relative to the most intense component in the plot.
Chromatogram scaled to the maximum height of the peak - To zoom out on a chromatogram, right-click the Chromatogram pane and select Zoom Out (if available). Otherwise, reset the chromatogram view to zoom out.
Reset chromatogram view to original scale
Procedure
- To reset the chromatogram view to the original scale, do either of the following:
- Right-click the Chromatogram pane and select Reset Scale.
- Double-click anywhere in the Chromatogram pane.
Copy chromatogram to the clipboard
Procedure
- To copy a chromatogram to the clipboard, right-click the Chromatogram pane and select Copy, Copy as Displayed, or Copy per Global Settings.
- The application copies the entire view of the Chromatogram pane to the clipboard, including all visible labeling and shading. You can then paste the copied picture into a third-party application file.
- See Copy and paste functions.
Display component peak labels
Procedure
- To display component peak labels in a chromatogram, right-click the Chromatogram pane, select Label, and select any of the following options (if available):
- Retention Time
- The application displays the retention time with one decimal digit on top of the peaks in all of the visible chromatograms.
- Peptide (or Oligonucleotide)
- The application displays the peptide (or oligonucleotide) information on top of the peaks in all of the visible chromatograms.
- For peptide mapping analysis and oligonucleotide analysis, an asterisk (*) appears at the end of the label if the sequence is modified.
- Alias
- The application displays the alias name of the component (if available).
- To copy the label data in the pane, see Copy all contents in a pane.