The following table describes the commands and parameters on the Queue page.
Parameter | Description |
|---|---|
Command bar | |
Stop/Pause/Run | (For the peptide mapping analysis, oligonucleotide analysis, and top down analysis workflows only) The Stop button cancels the processing of the currently running job and pauses the queue. The button changes to Run. (For the intact mass analysis workflow only) The Pause completes the processing of the currently running job and pauses the queue. The button changes to Run. Run resumes the processing of the remaining jobs in the queue. The button changes to Stop or Pause. |
Queue Manipulation | Contains commands that you use to remove jobs from the queue. |
Remove Selected | Removes the selected job or jobs from the queue. |
Remove Completed | Removes all completed jobs from the queue. |
Remove All | Removes all jobs from the queue. |
Open Results | Transfers you to the Process and Review page; enabled for completed jobs. |
Open Report | (For intact mass analysis only) Transfers you to the Reporting page to view the resulting report; enabled for completed jobs. |
Queue table | |
Record Number | Displays a sequential number that identifies each experiment in the queue. |
Experiment Name | Displays the name of the experiment. |
Status | Displays the real-time status of the analysis:
|
Experiment Type | Displays the type of experiment: Peptide Mapping, Oligonucleotide, Intact, or Top Down. |
Method Name | Displays the processing method used to run the experiment. |
Method Type | (For peptide mapping analysis only) Displays the type of peptide mapping method: Non Targeted or Targeted. |
Sequence Names | (Optional) Displays the name of the protein sequence or sequences assigned to the experiment. |
Raw File Names | Displays the name of the raw data file or files loaded for the experiment. |
Source Spectra Method | (For intact mass analysis only) Displays the method used to select the source spectra for deconvolution: Sliding Windows, Auto Peak Detection, or Average Over Selected Retention Time. |
Deconvolution Algorithm | (For intact mass analysis only) Displays the algorithm used for deconvolution: Xtract (Isotopically Resolved) or ReSpect (Isotopically Unresolved). |
Number of Chromatographic Peaks | (For intact mass analysis average over selected retention time deconvolution or auto peak detection experiments only) Displays the number of chromatographic peaks detected in the spectrum. If the application finds no chromatographic peaks, the value is 0. |
Number of Components Detected | (For intact mass analysis only) Displays the number of components detected from the deconvolution of each averaged spectrum from all found chromatographic peaks. If the application finds no chromatographic peaks, the value is 0. |
Submit Time | Displays the date and time that you placed the experiment into the run queue. |
Start Time | Displays the date and time that the processing started for the given experiment. |
Completion Time | Displays the date and time that the processing finished for the given experiment. |
Total Processing Time (min) | Displays the total time used to process the given experiment. |