Resume the paused queue - Resume the paused queue - BioPharma Finder 5.2 Oligonucleotide Analysis User Guide - BioPharma Finder Software - Resume the paused queue - BioPharma Finder Software - BioPharma Finder Software - topic
Ardia Platform > Software > BioPharma Finder Software
Desktop Software > BioPharma Finder Software
Document Type
User Guide
Software version
5.2
Procedure
Select the Queue tab to open the Queue page.
The paused queue shows jobs with the Submitted status that await processing.
Paused queue in the peptide mapping analysis workflow
Select Run in the command bar.
The Run button changes to a Stop button.
The application starts processing the first job with a Submitted status in the queue. After a few seconds of processing, the status of the job changes to Running.
When the processing of the experiment is completed, the application automatically continues to process subsequent jobs with a Submitted status in the queue.