When you submit an experiment for processing, the BioPharma Finder application adds the experiment to the run queue which can viewed on the Queue page.
When you submit multiple peptide mapping analysis, oligonucleotide analysis, intact mass analysis in automatic mode, or top down analysis jobs for processing, the application processes one job at a time.
TIP
While the application is processing jobs in the run queue, you can still create new processing methods or edit existing methods (even if the submitted jobs use those methods).
The application saves a copy of the processing method parameters when it saves the experiment, so subsequent method changes do not affect submitted or processing experiments.