Ardia Platform > Software > BioPharma Finder Software
Document Type
User Guide
Software version
5.2
Procedure
On the BioPharma Finder home page, select the Peptide Mapping Analysis, Oligonucleotide Analysis, Intact Mass Analysis, or Top Down Analysis workflow link in the Experiment Types pane or below the splash graphic.
The corresponding workflow page opens.
In the Processing Method table, select one or more checkboxes for the methods you want to export.
The Export Method button becomes active.
Active Export Method Button on the Intact Mass Analysis page
Select Export Method.
In the dialog box, browse to the appropriate folder where you want to save the method files.
Select OK.
A message appears confirming the export.
The exported processing methods are saved in the specified folder.