Delete raw data files - BioPharma Finder Software - Delete raw data files - BioPharma Finder Software - BioPharma Finder Software
BioPharma Finder 5.2 Oligonucleotide Analysis User Guide
- Product Tree
- Ardia Platform > Software > BioPharma Finder Software
- Desktop Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- On the BioPharma Finder home page, select the Peptide Mapping Analysis, Oligonucleotide Analysis, Intact Mass Analysis, or Top Down Analysis workflow link in the Experiment Types pane or below the splash graphic.
- The corresponding workflow page opens.
- In the list of raw data files in the Load Raw Data pane, select the name of the raw data file you want to remove.
- You can select only one raw data file at a time.
- The application highlights the selected file in blue.
- Press the DELETE key.
- When prompted, select Yes in the confirmation box to proceed.
- The application deletes the selected raw data file from the experiment.
- If the deleted file was assigned as the reference condition for the experiment, reassign the reference condition as appropriate.