Purge all BioPharma Finder files - Purge all BioPharma Finder files - BioPharma Finder 5.2 Oligonucleotide Analysis User Guide - BioPharma Finder Software - Purge all BioPharma Finder files - BioPharma Finder Software - BioPharma Finder Software - topic
BioPharma Finder 5.2 Oligonucleotide Analysis User Guide
- Product Tree
- Ardia Platform > Software > BioPharma Finder Software
- Desktop Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- On the BioPharma Finder home page, select Help > Ardia Platform Management > Data Space Optimization > Clear All.
- When prompted, select Yes to continue.