In addition to the Process and Review page, you can view the results of your oligonucleotide analysis experiment on the Modification Summary page. The Modification Summary page provides detailed information regarding the recovery status and abundance of all detected modifications.
Procedure
- Open the results of your experiment from the Queue page or from the Load Results page.
- See Open results.
- The Process and Review page opens by default.
- In the navigation bar, select the Mapping tab.
- By default, the application opens the Coverage page under the Mapping tab.
- In addition to the Coverage page, the Mapping tab also contains the Modification Summary page.
- On the Mapping page, select the Modification Summary subtab.
- The Modification Summary page opens, displaying the Modification Results and Components tables on the left side and the Modification Plot on the right side.
Modification Summary page