The Trend MS Area pane on the Process and Review page displays bar plots representing the MS Area values for each condition (raw data file) for a selected component. The Trend Ratio pane is visible only for experiments with multiple raw data files.
Procedure
  1. Open the results of your experiment from the Queue page or from the Load Results page.
  2. See Open results.
  3. The Process and Review page opens by default and displays the results of your oligonucleotide analysis experiment, including the Trend MS Area plots for experiments with multiple raw data files.
  4. On the left side of the Process and Review page, select the Trend MS Area tab.
  5. Select the row for a component (or a specific raw data file under a component) in the Results table.
  6. The Trend MS Area plot shows the MS Area value for each condition (raw data file) used in the experiment, with each condition depicted in a different color.
  7. The values in the plot come from the associated column in the Results table.
  8. The order of the conditions underneath the bars in the plot is the same order as the conditions entered on the Oligonucleotide Analysis page.
Trend MS Area pane
Trend MS Area pane