When you submit an oligonucleotide analysis experiment on the Oligonucleotide Analysis page, the application submits the experiment to the run queue and opens the Queue page. The Queue page lists all of the submitted, in-progress, completed, and cancelled jobs in the run queue.
- When your experiment is actively processing, its job status shows Running.
- When processing is complete, its job status shows Completed.
- If you submit your experiment while another experiment is processing, your experiment is added to the run queue and its job status shows Submitted.
- Other possible statuses include Cancelled, Abort, and Error.
For more information on managing the run queue, see Use the run queue.
The following table describes the columns on the Queue page.
Column | Description |
|---|---|
Record Number | Record number of the current experiment |
Experiment Name | Name of the current experiment |
Status | Status of the job: Submitted, Running, Cancelled, Completed, Abort, or Error |
Experiment Type | Experiment type selected |
Method Name | Name of the processing method selected |
Method Type | Method type (non-targeted or targeted) selected Note: Currently only non-targeted methods are supported for oligonucleotide analysis experiments. |
Sequence Name | Name of the oligonucleotide sequence selected (if applicable) |
Raw File Names | Names of raw data files uploaded |
Submit Time | Time when you submitted the experiment for processing |
Start Time | Time when the application started processing the experiment |
Completion Time | Time when the application completed processing the experiment |
Processing Time (min) | Time taken to process the experiment from start time to completion time |