Use the oligonucleotide analysis workflow in the BioPharma Finder application to perform a complete characterization of nucleotide-based pharmaceuticals. This platform provides a high-confidence analysis for the fully automated characterization of oligonucleotides. It performs component detection, nucleotide identification, and identification and quantification of post-translational modifications (PTMs) using data collected on Thermo Scientificâ„¢ instruments.