The following table describes the types of information in the table on the Workbook Editor page for an intact workbook.
Column | Description |
|---|---|
Component level | |
+/− | Shows or hides the lower level of charge state information related to the current component row. |
Row number (bolded) | The number assigned to each visible workbook row in the table. This sequential numbering does not change when you sort or filter the table. |
(Check box) | Selected checkbox indicates workbook entry for export. To select or clear all of the checkboxes at once, select or clear the checkbox in the column header. If you filter the table, the following occurs:
|
Level | Indicates that the row is displaying component information (top level). |
(Various component information columns) | |
Charge state level | |
Row number (bolded) | The number assigned to each visible charge state row in the table. This sequential numbering does not change when you sort or filter the table. |
Level | Indicates that the row is displaying charge state information (lower level). |