On the Load Results page, you can manage experiment results in the Master List.
Delete the results of one or more experiments
Procedure
- Select the checkbox for one or more experiment rows.
- Select Delete in the command bar.
- The experiments are removed from the Master List.
NOTE
The action permanently deletes the selected results from the BioPharma Finder database. If the selected experiments still exist in the run queue on the Queue page, the application removes them from the queue as well.
Import the results of an experiment
Procedure
- Select Import Results on the command bar.
- In the dialog box, select one or more BioPharma Finder results files.
- Peptide mapping analysis results files appear as "filename_peptide.resultsbpf".
- Oligonucleotide analysis results files appear as "filename_oligonucleotide.resultsbpf".
- Intact mass analysis results files appear as "filename_intact.resultsbpf".
- Top down analysis results files appear as "filename_topdown.resultsbpf".
NOTE
If you are importing multiple results files simultaneously, the files must be in the same folder.
- Select Open.
- A message appears confirming the import.
- The imported result(s) appear in the Master List.
Export the results of an experiment
Procedure
- Select the checkbox for one or more experiment rows in the Working List.
- Do one of the following:
- Select Export Results > Export experiment with raw files on the command bar.
- Select Export Results > Export experiment without raw files on the command bar.
NOTE
You can only view the experiment results when you include the raw files in the export. Additionally, you must save the raw data file in the same directory as the processed data file in order to view the spectra in the experiment.
NOTE
If you export multiple experiments with their raw files and the total file size exceeds 5 GB, an error message appears and the experiments are not exported with the raw files. In the error message dialog box, select OK to export the experiments without the raw files.
- In the dialog box, browse to the appropriate folder location and select OK.
- A message appears confirming the export. The application saves the exported experiment result in the specified folder.
- Peptide mapping analysis results files appear as "filename_peptide.resultsbpf".
- Oligonucleotide analysis results files appear as "filename_oligonucleotide.resultsbpf".
- Intact mass analysis results files appear as "filename_intact.resultsbpf".
- Top down analysis results files appear as "filename_topdown.resultsbpf".