You can select a different identification for the selected component in the Matched Sequence pane on the Process and Review page.
Prerequisites
- You have loaded the experiment results and are viewing the Matched Sequence pane on the Process and Review page.
Procedure
- On the Process and Review page, in the Results table, select the row of a matched component.
- In the Target Match Sequence table in the Matched Sequence pane, select the checkbox for a specific identification row.
- See Target Match Sequence table parameters.
- The new selection overwrites the default best matched sequence identification.
- Selecting a new identification automatically updates the identification results for the component in the Results table and the corresponding peak label in the Deconvoluted Spectrum on the Process and Review page.
- (Optional) After you make modifications to the identifications, select Save Results As in the command bar of the Process and Review page to save to a new experiment or overwrite the current experiment results.