Procedure
- On the BioPharma Finder home page, select Intact Mass Analysis in the Experiment Types navigation pane.
- Select the Load Results tab on the navigation bar.
- The Load Results page opens and a table displays all of the previously saved intact mass analysis results, listed in chronological order of completion. You can sort and/or filter the table as needed.
Load Results table columns - The Load Results table displays the following information:
- •Experiment name
- •Processing method name
- •Raw data file names/file paths
- •Source spectra (deconvolution) method
- •Deconvolution algorithm
- •Number of chromatographic peaks and detected components in the analysis
- •Time of completion and processing time
- To open the results of an experiment, do one of the following:
- Double-click the experiment row.
- Select the experiment row and select Load Results on the command bar.
- The Process and Review page opens by default. You can also select the Reporting tab to view the deconvolution report for the experiment and the Spectra Comparison tab to generate a mirror plot of two deconvolved spectra.
NOTE
For more information on organizing experiment results using the Experiment Manager on the Load Results page, see Organize experiment results.