For intact mass experiments processed in Automatic mode, you can open the results of the experiment directly from the run queue. For experiments processed and saved in Manual mode, see Open results from the Load Results page.
Procedure
  1. On the BioPharma Finder home page, select Intact Mass Analysis in the Experiment Types navigation pane (if necessary).
  2. Select the Queue tab on the navigation bar.
  3. The Queue page opens and displays all saved, completed, running, submitted, and canceled jobs in the run queue.
  4. NOTE
    You cannot open experiment results if the application has not completely processed the experiment. The Open Results button is inactive until the selected job is completed.
    Opening experiment results does not interfere with the application processing subsequent experiments in the run queue.
  5. In the run queue, do one of the following:
  6. Double-click the row of a completed job.
  7. Select the row of a completed job and then select Open Results in the command bar.
  8. The Process and Review page opens by default. You can also select the Reporting tab to view the deconvolution report for the experiment and the Spectra Comparison tab to generate a mirror plot of two deconvolved spectra.
NOTE
If you delete a job in the run queue on the Queue page, you can no longer open the experiment results from the Queue page. However, the BioPharma Finder application saves all completed jobs; results can be opened from the Load Results page even after they are deleted from the run queue.