You can replace the bases, backbone linkers, and 2' ribose subunits of an oligonucleotide sequence with custom building blocks or add custom chemical groups to the 5' and 3' terminals of the sequence chains using the Oligonucleotide Sequence Editor. For information on how to create and manage custom building blocks, see Manage custom modifications and building blocks for oligonucleotides.
Procedure
- In the Oligonucleotide Sequence Editor, select the title bar for Assign Variable Modifications.
- Oligo Sequence Map area displays all the oligo building blocks comprising the sequence in read-only mode.
- On the right side of the Oligonucleotide Sequence Editor, select the Edit Sequence title bar to open the Edit Sequence pane.
Edit Sequence pane showing Select Chain options - To edit a specific chain, select the chain number that you want modify in the Select Chain drop-down list.
Custom building block options for each triplet in a chain - Select a 5'-terminal for the chain from the drop-down list above the table.
- In the table, use the drop-down lists to specify the three building blocks (backbone linker, base, and 2'-ribose) for each triplet in the sequence.
- Select a 3'-terminal for the chain from the drop-down list below the table.
- Select Apply.
- The updated oligo building blocks appear in the chain in the Oligo Sequence Map pane.
- The application displays the recalculated masses and the revised formula in the Sequence Information.
- To edit all of the chains in the sequence, select the All Chains option in the Select Chain drop-down list and then do the following:
Subunit, To, and From selections in the Edit Sequence pane - Select a subunit from the Subunit drop-down list.
- Select the building block or modification you want to change from the From drop-down list.
- Select the building block or modification you want to add from the To drop-down list.
- Select Apply.
- All subunits of the specified type with be globally replaced with the custom building block.
- The updated oligo building blocks appear in the chain in the Oligo Sequence Map pane.
- The application displays the recalculated masses and the revised formula in the Sequence Information.
Oligo Sequence Map showing oligonucleotide sequence modified with custom building blocks - Select Save or Save as New in the command bar to save the modifications. Or, if you do not want to save the modifications to the sequence, select Cancel.
- See Save an oligonucleotide sequence.