You can open experiment results directly from the run queue on the Queue page (for Automatic mode experiments) or from the Load Results page (for both Automatic and Manual mode experiments).
  • The Queue page displays the run queue containing all of the saved, submitted, running, completed, and canceled jobs. After a job is completed, you can open the experiment results from the Queue page to view the processed data.
  • On the Load Results page of the intact mass analysis workflow, you can open, delete, import, and export any previously saved experiment results.