When you submit an intact mass analysis experiment in Automatic mode on the Intact Mass Analysis page, the application submits the experiment to the run queue and opens the Queue page. The Queue page lists all of the submitted jobs, in-progress jobs, completed jobs, and cancelled jobs in the run queue.
- When your experiment is actively processing, its job status shows Running.
- When processing is complete, its job status shows Completed.
- If you submit your experiment while another experiment is processing, your experiment is added to the run queue and its job status shows Submitted.
- Other possible statuses include Cancelled, Abort, and Error.
For more information on managing the run queue, see Use the run queue.
The following table describes the columns on the Queue page.
Column | Description |
|---|---|
Record Number | Record number of the current experiment |
Experiment Name | Name of the current experiment |
Status | Status of the job: Submitted, Running, Cancelled, Completed, Abort, or Error |
Number of Chromatographic Peaks | Number of peaks detected in the chromatogram |
Number of Detected Components | Number of components detected in the deconvolved spectrum |
Experiment Type | Experiment type selected (Intact) |
Method Name | Name of the processing method selected |
Sequence Names | Name of the protein or oligonucleotide sequence(s) selected (if applicable) |
Raw File Names | Names of the raw data files uploaded |
Source Spectra Method | Source spectra (deconvolution) method used in the selected processing method: Average Over Selected Retention Time, Auto Peak Detection, or Sliding Windows |
Deconvolution Algorithm | Deconvolution algorithm used in the selected processing method: Xtract of ReSpect |
Submit Time | Time when you submitted the experiment for processing |
Start Time | Time when the application started processing the experiment |
Completion Time | Time when the application completed processing the experiment |
Processing Time (min) | Time taken to process the experiment from start time to completion time |