The Source Spectrum Evidence Plot section of a report displays a source spectrum plot for each component in the sample. This section appears only if you select the Component Source of Evidence Plots option on the Parameters > Report page in the processing method editing wizard. The plot shows the peaks or isotopic clusters in the scan that are associated with a particular component. See View the source spectra.
The following figure shows the Source Spectrum Evidence Plot for an Xtract deconvolution.
Source Spectrum Evidence Plot section for an Xtract deconvolution
image/svg+xml
The following figure shows the Source Spectrum Evidence Plot for a ReSpect deconvolution.
Source Spectrum Evidence Plot section for a ReSpect deconvolution
image/svg+xml
The following table lists the parameters for the source spectrum shown in the Source Spectrum Evidence Plot section.
Source Spectrum Evidence Plot section parameters
Parameter
Description
Relative Intensity (y axis)
Displays the ratio of the intensity of a specific peak to the intensity of the peak with the highest intensity.
m/z (x axis)
Displays the mass-to-charge ratio of ions formed from molecules.