The Component Detail Tables section of a report displays a table for each component in the sample. This section appears only if you select the Component Detail Tables option on the Parameters > Report page in the processing method editing wizard. The Component Details tables show all the charge states that the BioPharma Finder application detected for a particular component. It displays the same parameters as those displayed in the Results table on the Process and Review page and these parameters depend on the type of deconvolution used during processing. See Results table parameters (for single file or batch format experiments) and Results table parameters (multiconsensus format experiments).
The following figure shows the Component Details Tables section for a single/auto peak detection experiment using Xtract deconvolution.
The following figure shows the Component Details Tables section for a single/auto peak detection experiment using ReSpect deconvolution.
The following figure shows the Component Details Tables section for a sliding windows experiment using Xtract deconvolution.
The following figure shows the Component Details Tables section for a sliding windows experiment using ReSpect deconvolution.