The Deconvoluted Spectrum section of a report, shown in the following figure, displays the same spectrum that appears in the Deconvoluted Spectrum pane of the Process and Review page. See View the deconvoluted spectra.
Deconvoluted Spectrum section
Deconvoluted Spectrum section
The following table lists the parameters in the Deconvoluted Spectrum section.
Deconvoluted Spectrum section parameters
Parameter
Description
Relative Intensity (y axis)
Displays the ratio of the intensity of a specific peak to the intensity of the peak with the highest intensity.
Mass (x axis)
Displays the mass of the ions formed from molecules.