The Source Spectrum section of a report, shown in the following figure, displays the spectrum in the Source Spectrum pane of the Parameters > Component Detection page in the processing method editing wizard or the Process and Review page. See View the source spectra. The report does not include this section for experiments using sliding windows deconvolution because there is no single source spectrum for the results.
Source Spectrum section
Source Spectrum section
The following table lists the parameters in the Source Spectrum section.
Source Spectrum section parameters
Parameter
Description
Relative Abundance (y axis)
Displays the ratio of the abundance of a specific peak to the abundance of the peak with the highest abundance.
Mass or m/z (x axis)
Displays the mass or mass-to-charge ratio of the ions formed from molecules.