Right-clicking in the Average DAR pane on the Process and Review page opens a shortcut menu that contains the command listed in the following table.
Command | Description |
|---|---|
Export Component Specific Summary | Exports the values in the Component Specific Summary table to a Microsoft™ Excel™ workbook (.xlsx). The default file name is Experiment Name_DAR_ComponentSummary.xls. If you do not browse to a different folder, the application places the exported file in the folder with the raw data files for the experiment. |