Right-clicking the Deconvoluted Spectrum pane on the Process and Review page opens a shortcut menu that contains the commands listed in the following table.
Command | Description |
|---|---|
Reset Scale | Restores the original scale to the pane. |
Copy as Displayed | Copies the image in the pane to the clipboard as currently displayed. |
Copy per Global Settings | Copies the image in the pane to the clipboard based on the global image dimensions set in the Intact Mass Analysis Settings dialog box. |
Copy Data | Copies mass data (x axis) and intensity data (y axis) from the pane to the clipboard so that you can paste it into a Microsoft™ Excel™ spreadsheet or another application. For Xtract deconvolution, the copied data consists of a centroid spectrum. For ReSpect deconvolution, the copied data consists of a profile spectrum. |
Zoom Out | Shrinks the view in the pane by a factor of 2. |
Zoom In | Enlarges the view in the pane by a factor of 2. |
Show Identification Markers (checkbox) | Turns on and off to show or hide the markers for identified peaks. Active only for target sequence matching experiments. Default: On (When you close and reopen the application, the identification markers automatically appear.) |
Identification Markers Color | Changes the color of the identification markers. Active only when the Show Identification Markers command is turned on. Select a different color from the palette, or select Advanced to enter specific RGB, HSL, or CMYK values for the new color. Default: Orange (When you close and reopen the application, the color you previously selected reverts back to orange.) |