Right-click the Results table on the Process and Review page to open a shortcut menu with the commands listed in the following table.
Command | Description |
|---|---|
Set As Reference Component | Sets the mass of the chosen component as the reference mass for the Results table. |
Export All | Exports all data in the Results table to a Microsoft™ Excel™ workbook (.xlsx). |
Export Checked | Exports only the selected data in the Results table to a Microsoft™ Excel™ workbook (.xlsx). |
Export Json | Exports all results in the Results table to a text-based JavaScript file for data interchange. |
Component Level Only | Exports only the component-level results in the table. The exported data does not include the results at the raw data file and charge state levels. |
All Levels | Exports the results at all levels in the table. |
Save As Intact Workbook | (For average over selected retention time or sliding windows experiments) Saves all or selected component results and associated processing parameters to a workbook (.wbpf) that is Chromeleon™-compatible and used for targeted experiments. |