An intact workbook contains the processing parameters and intact deconvolution results for an intact mass analysis experiment. You can save all or selected results for use in the Chromeleon™ chromatography data system.
NOTE
You can save results to an intact workbook only for average over retention time or sliding windows experiments, not for auto peak detection experiments.
Prerequisites
- You have loaded the experiment results are viewing the Process and Review page.
Procedure
- On the Process and Review page, select the checkbox in the row of each component in the Results table that you want to save to a workbook.
TIP
To select all of the rows, select the checkbox in the column header.
- Right-click anywhere in the Results table and select Save As Intact Workbook from the shortcut menu.
- Select one of the following submenu commands:
- All to export all results to a workbook.
- Checked to export only the selected results to a workbook.
- The Save Intact Workbook As dialog box opens.
- See Results table commands.
- In the Save Intact Workbook As dialog box, type a new name in the Workbook Name box to create a new workbook or keep the same name to overwrite the workbook if it already exists.
- The default workbook name is the same as the experiment name.
- Use only alphanumeric, space, underscore "_", and period "." characters in the workbook name.
- (Optional) In the Description box, type a description for the workbook.
- Select Save.
- If the workbook is currently open for editing on the Workbook Editor page, indicate whether you want the application to automatically close the open workbook before saving.
- After saving, you can manage the workbook using the Intact Workbook page.
- See Use a Chromeleon-compatible workbook: Intact.