You can export all or selected results from the Results table to a Microsoft™ Excel™ workbook (.xlsx) or Json file.
Prerequisites
- You have loaded the experiment results are viewing the Process and Review page.
Procedure
- On the Process and Review page, right-click anywhere in the Results table. Select one of the following menu commands:
- Export All to export all results of a certain type to a Microsoft™ Excel™ workbook (.xlsx).
- Export Checked to export only the selected results of a certain type to a Microsoft™ Excel™ workbook (.xlsx).
- Export Json to export all results to a text-based JavaScript file for data interchange.
TIP
To select an entire row of results to export, select the checkbox in that row.
To select all of the rows, select the checkbox in the column header.
- For the Export All or Export Checked commands, select one of the following submenu commands:
- Component Levels Only to export only the results at the component level in the table.
- The exported data does not include any results at the raw data file and charge state levels.
- All Levels to export the results at all levels.
- See Results table commands.
- In the Save As dialog box, browse to the file location where you want to store the exported results.
- By default, the file name is the same as the experiment name.
- Select Save.