Right-clicking the Mirror Plot pane on the Spectra Comparison page opens a shortcut menu that contains the commands listed in the following table.
Command | Description |
|---|---|
Reverse Order | Reverses the order of the two spectra in the Mirror Plot pane. |
Reset Scale | Restores the original scale to the Mirror Plot pane. |
Copy as Displayed | Copies the image in the Mirror Plot pane to the clipboard. |
Copy per Global Settings | Copies the image in the Mirror Plot pane to the clipboard based on the global image dimensions set in the Intact Mass Analysis Settings dialog box. |
Zoom Out | Shrinks the view in the pane by a factor of 2. |
Zoom In | Enlarges the view in the pane by a factor of 2. |