You can open the results of your intact mass analysis experiment from either the Queue or Load Results page.
Intact mass analysis results are displayed on the Process and Review page. In addition to viewing your results, you can quickly change processing parameters and resubmit your job using the real-time optimization (RTO) feature on the Process and Review page.
Additionally, you can view the deconvolution report for your experiment on the Reporting page and can compare two deconvoluted spectra by generating a mirror plot on the Spectra Comparison page.