The following table describes the parameters on the Report page.
Parameter | Description |
|---|---|
Reporting Parameters area Select the checkbox for each report section you want to generate and display in the report. | |
Sample Information | Generates the Sample Information section of the report. |
Chromatogram with Parameters | Generates the Chromatogram Parameters and Chromatogram sections of the report. |
Source Spectrum | Generates the Source Spectrum section of the report. |
Deconvolution Parameters | Generates the Main Parameters, Advanced Parameters, Source Spectra Parameters, and Sequences Information sections of the report. |
Deconvoluted Spectrum | Generates the Deconvoluted Spectrum section of the report. |
Components Table | Generates the Masses Table section of the report, which displays information from the Results table on the Process and Review page. |
Component Detail Tables | Generates the Monoisotopic Mass table (for Xtract) or Average Mass table (for ReSpect) for each component in the Component Detail Tables section of the report. |
Component Source of Evidence Plots | Generates the Source Spectrum Evidence Plot section for each component in the Component Detail Tables section of the report. |
Automation Parameters area Select the checkbox for each report section you want to generate and display in the report. The parameters in this pane apply to Automatic mode experiments only. | |
Save to PDF | When selected (default), saves the generated deconvolution report (as a .pdf file) in the same folder as the raw data files, using the following file name format: RawFileName_ExperimentName.pdf |
Auto Print | When selected, automatically prints the generated deconvolution report. When you select this option, the Select a Printer box becomes available so that you can select a printer. |
Save to .jpg | When selected (default), saves the chromatographic peak data to both a .jpg file and .csv file. When cleared, saves chromatographic peak data only to a .csv file. For an average over selected retention time deconvolution/auto peak detection experiment, the application saves each peak to an individual file. For a sliding windows deconvolution experiment, it saves all of the peaks to one file. The file names have the following format: RawFileName_ExperimentName_ScanRange_DateStamp_TimeStamp_[AM|PM]_[XT|RSP]_AUTO.jpg RawFileName_ExperimentName_ScanRange_DateStamp_TimeStamp_[AM|PM]_[XT|RSP]_AUTO.csv where XT refers to Xtract and RSP refers to ReSpect. |
Concatenate All Reports | When selected (default), combines all reports for all deconvoluted spectra from the same raw data file into one deconvolution report (.pdf file). When cleared, creates a deconvolution report file (.pdf file) for each chromatographic peak in the raw data file. |
Select a Printer | Displays a list of available printers that can print the deconvolution report (.pdf file). This option becomes available when you select the Auto Print option. |