On the Component Detection page, the left side of the page displays the editable parameters related to component detection in your intact mass analysis experiment. These parameters vary, depending on the deconvolution algorithm (Xtract or ReSpect) and source spectra (deconvolution) method (average over selected retention time, automatic peak detection, or sliding windows) that you select.
Refer to the following topics for more information on the parameters displayed in each section of the Component Detection page:
- Chromatogram Parameters area: Chromatogram Parameters area parameters
- Source Spectra Method area: Source Spectra Method area parameters
- Deconvolution Algorithm area: Xtract deconvolution parameters and ReSpect deconvolution parameters