After you add a sequence (with the "Intact Protein" category for proteins or the "Intact Deconvolution" category for oligonucleotides) to the Sequence Manager, you can start an intact mass analysis experiment by selecting Intact Mass Analysis on the BioPharma Finder home page.
You can run an intact mass analysis experiment in Manual or Automatic mode. See Processing modes: Manual vs. Automatic.