Save an oligonucleotide sequence under a new name - Save an oligonucleotide sequence under a new name - BioPharma Finder 5.2 Intact Mass Analysis User Guide - BioPharma Finder Software - Save an oligonucleotide sequence under a new name - BioPharma Finder Software - BioPharma Finder Software - topic
BioPharma Finder 5.2 Intact Mass Analysis User Guide
- Product Tree
- Ardia Platform > Software > BioPharma Finder Software
- Desktop Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- In the Oligonucleotide Editor, select Save As New on the command bar.
- The Save As New dialog box opens.
- In the New Sequence Name box, type the new name of the oligonucleotide sequence.
- (Optional) In the Description box, type a description of the oligonucleotide sequence.
- Select OK.
- Information from the new saved sequence populates the columns of the sequence table on the Sequence Manager page.