Upon installation, the BioPharma Finder application provides a default list of variable modifications for oligonucleotide sequences. You can access this list from the Oligonucleotide Sequence Editor and define which modifications appear in the list of variable modifications in the Assign Variable Modifications pane and which modifications are selected for quick loading.